Assay Design



Gene Transcripts

Gene Symbol
guca1a
Entrez Gene ID
779962
Gene Name
guanylate cyclase activator 1A (retina)
Gene Aliases
-
Chromosome Location
Chr.3: 80303486 - 80314839 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079039.1 NP_001072507.1 3 - 4 921 89
GenBank mRNA BC121947.1 - 3 - 4 921 89

View Full Product Details