Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079083.1 | NP_001072551.1 | 11 - 12 | 1022 | 87 |
XM_012957278.2 | XP_012812732.1 | 10 - 11 | 1115 | 87 | |
XM_018091349.1 | XP_017946838.1 | 9 - 10 | 846 | 87 | |
GenBank mRNA | BC122064.1 | - | 11 - 12 | 1022 | 87 |