Assay Design



Gene Transcripts

Gene Symbol
b3glct
Entrez Gene ID
780006
Gene Name
beta 3-glucosyltransferase
Gene Aliases
b3galtl
Chromosome Location
Chr.2: 150789562 - 150976728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079083.1 NP_001072551.1 1 - 2 103 67
GenBank mRNA BC122064.1 - 1 - 2 103 67

View Full Product Details