Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079083.1 | NP_001072551.1 | 4 - 5 | 313 | 85 |
XM_012957278.2 | XP_012812732.1 | 3 - 4 | 406 | 85 | |
XM_018091349.1 | XP_017946838.1 | 1 - 2 | 84 | 85 | |
GenBank mRNA | BC122064.1 | - | 4 - 5 | 313 | 85 |