Assay Design



Gene Transcripts

Gene Symbol
duoxa1
Entrez Gene ID
780018
Gene Name
dual oxidase maturation factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 85957966 - 85969891 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079095.1 NP_001072563.1 3 - 4 258 115
GenBank mRNA BC122088.1 - 3 - 4 258 115

View Full Product Details