Assay Design



Gene Transcripts

Gene Symbol
nkain1
Entrez Gene ID
780051
Gene Name
Sodium/potassium transporting ATPase interacting 1cting 1
Gene Aliases
fam77c
Chromosome Location
Chr.2: 53379816 - 53398822 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079128.1 NP_001072596.1 4 - 5 695 97
GenBank mRNA BC122964.1 - 4 - 5 695 97
BC135653.1 - 4 - 5 687 97

View Full Product Details