Assay Design



Gene Transcripts

Gene Symbol
galnt4
Entrez Gene ID
780162
Gene Name
polypeptide N-acetylgalactosaminyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.6: 38278397 - 38307476 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079237.2 NP_001072705.1 4 - 5 726 97
GenBank mRNA BC124051.1 - 3 - 4 643 97
BC135911.1 - 4 - 5 726 97

View Full Product Details