Assay Design



Gene Transcripts

Gene Symbol
fbxo22
Entrez Gene ID
780359
Gene Name
F-box protein 22
Gene Aliases
-
Chromosome Location
Chr.3: 80527336 - 80542249 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079429.1 NP_001072897.1 3 - 4 420 58
XM_018092186.1 XP_017947675.1 3 - 4 312 58
GenBank mRNA BC118853.1 - 3 - 4 420 58
BC135261.1 - 3 - 4 411 58

View Full Product Details