Assay Design



Gene Transcripts

Gene Symbol
bnip1
Entrez Gene ID
780363
Gene Name
BCL2/adenovirus E1B 19kDa interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 33917339 - 33925413 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079433.1 NP_001072901.1 1 - 2 115 93
GenBank mRNA BC121430.1 - 1 - 2 115 93

View Full Product Details