Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079438.2 | NP_001072906.2 | 2 - 3 | 445 | 98 |
XM_012961436.2 | XP_012816890.1 | 2 - 3 | 1265 | 98 | |
GenBank mRNA | BC121975.1 | - | 1 - 2 | 900 | 98 |
BC136031.1 | - | 2 - 3 | 445 | 98 |