Assay Design



Gene Transcripts

Gene Symbol
bmt2
Entrez Gene ID
496879
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Gene Aliases
c7orf60
Chromosome Location
Chr.3: 57274732 - 57285120 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083350.2 NP_001076819.1 3 - 4 512 101
XM_012958947.2 XP_012814401.1 3 - 4 666 101
XM_018092060.1 XP_017947549.1 3 - 4 356 101
GenBank mRNA BC088770.1 - 3 - 4 451 101
BC135938.1 - 3 - 4 464 101
DN093113.1 - 3 - 4 512 101

View Full Product Details