Assay Design



Gene Transcripts

Gene Symbol
lmnb3
Entrez Gene ID
733537
Gene Name
lamin B3
Gene Aliases
LB3, lamin-b3
Chromosome Location
Chr.3: 95699929 - 95717744 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083354.2 NP_001076823.1 8 - 9 1546 90
XM_012959159.2 XP_012814613.1 8 - 9 2075 90
GenBank mRNA BC135750.1 - 8 - 9 1546 90

View Full Product Details