Assay Design



Gene Transcripts

Gene Symbol
cmas
Entrez Gene ID
100036739
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097281.1 NP_001090750.1 - 582 92
GenBank mRNA AM262832.1 - - 582 92

View Full Product Details