Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097320.1 | NP_001090789.1 | 3 - 4 | 393 | 88 |
XM_012957609.2 | XP_012813063.1 | 4 - 5 | 452 | 88 | |
XM_018091169.1 | XP_017946658.1 | 4 - 5 | 452 | 88 | |
XM_018091179.1 | XP_017946668.1 | 3 - 4 | 930 | 88 | |
XM_018091197.1 | XP_017946686.1 | 4 - 5 | 692 | 88 | |
XM_018091214.1 | XP_017946703.1 | 3 - 4 | 595 | 88 | |
XM_018091227.1 | XP_017946716.1 | 3 - 4 | 617 | 88 | |
XM_018091240.1 | XP_017946729.1 | 3 - 4 | 526 | 88 | |
GenBank mRNA | BC135179.1 | - | 3 - 4 | 393 | 88 |