Assay Design



Gene Transcripts

Gene Symbol
rnls
Entrez Gene ID
100037883
Gene Name
renalase, FAD-dependent amine oxidase
Gene Aliases
-
Chromosome Location
Chr.7: 4556303 - 4603892 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097322.1 NP_001090791.1 6 - 7 887 92
XM_012965827.2 XP_012821281.1 6 - 7 1001 92
GenBank mRNA BC135183.1 - 6 - 7 887 92

View Full Product Details