Assay Design



Gene Transcripts

Gene Symbol
agpat1
Entrez Gene ID
100037898
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-agpat1, g15, lpaat-alpha, lpaata
Chromosome Location
Chr.8: 54298648 - 54329404 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097333.1 NP_001090802.1 8 - 9 995 99
XM_012967818.2 XP_012823272.1 9 - 10 984 99
XM_012967820.2 XP_012823274.1 8 - 9 912 99
XM_012967821.2 XP_012823275.1 8 - 9 860 99
XM_012967822.2 XP_012823276.1 9 - 10 926 99
XM_012967823.2 XP_012823277.1 9 - 10 985 99
XM_012967824.2 XP_012823278.1 9 - 10 874 99
XM_012967825.2 XP_012823279.1 8 - 9 841 99
XM_012967826.2 XP_012823280.1 7 - 8 788 99
XM_012967827.2 XP_012823281.1 8 - 9 902 99
XM_018096082.1 XP_017951571.1 9 - 10 874 99
XM_018096083.1 XP_017951572.1 7 - 8 973 99
XM_018096084.1 XP_017951573.1 7 - 8 764 99
GenBank mRNA BC135226.1 - 8 - 9 995 99

View Full Product Details