Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097408.1 | NP_001090877.1 | 1 - 2 | 160 | 58 |
XM_018095211.1 | XP_017950700.1 | 1 - 2 | 201 | 58 | |
GenBank mRNA | BC135334.1 | - | 1 - 2 | 160 | 58 |
CT030416.1 | - | 1 - 2 | 97 | 58 |