Assay Design



Gene Transcripts

Gene Symbol
mydgf
Entrez Gene ID
100101683
Gene Name
myeloid-derived growth factor
Gene Aliases
c19orf10, il25, il27, il27w, r33729_1, sf20
Chromosome Location
Chr.1: 74711897 - 74717101 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100209.1 NP_001093679.1 5 - 6 455 89
GenBank mRNA BC135669.1 - 5 - 6 455 89

View Full Product Details