Assay Design



Gene Transcripts

Gene Symbol
lhx1
Entrez Gene ID
100101708
Gene Name
LIM homeobox 1
Gene Aliases
Xlim-1, Xlim1, lim-1, lim1
Chromosome Location
Chr.2: 49856639 - 49867517 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100228.1 NP_001093698.1 2 - 3 1023 72
GenBank mRNA BC135731.1 - 2 - 3 1023 72

View Full Product Details