Assay Design



Gene Transcripts

Gene Symbol
smn2
Entrez Gene ID
100101724
Gene Name
survival of motor neuron 2, centromeric
Gene Aliases
bcd541, c-bcd541, gemin-1, smnc
Chromosome Location
Chr.1: 167203861 - 167210962 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100240.1 NP_001093710.1 7 - 8 844 90
XM_018092087.1 XP_017947576.1 6 - 7 737 90
GenBank mRNA BC135221.1 - 7 - 8 844 90

View Full Product Details