Assay Design



Gene Transcripts

Gene Symbol
olfm1
Entrez Gene ID
100101729
Gene Name
olfactomedin 1
Gene Aliases
amy, noe1, noelin-1, noelin1, olfa
Chromosome Location
Chr.Un|NW_016683374: 1269189 - 1301888 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100244.1 NP_001093714.1 - 450 56
GenBank mRNA BC135743.1 - - 450 56

View Full Product Details