Assay Design



Gene Transcripts

Gene Symbol
ypel2
Entrez Gene ID
100124702
Gene Name
yippee like 2
Gene Aliases
fksg4
Chromosome Location
Chr.2: 87025740 - 87069819 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102689.1 NP_001096159.1 3 - 4 508 64
GenBank mRNA BC129024.1 - 3 - 4 508 64

View Full Product Details