Assay Design



Gene Transcripts

Gene Symbol
tcta
Entrez Gene ID
100124829
Gene Name
T cell leukemia translocation altered
Gene Aliases
-
Chromosome Location
Chr.4: 115505571 - 115511955 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102794.2 NP_001096264.1 2 - 3 471 77
GenBank mRNA BC135472.1 - 2 - 3 395 77
BC155508.1 - 2 - 3 360 77
CN094320.1 - 2 - 3 475 77
ES679293.1 - 2 - 3 465 77

View Full Product Details