Assay Design



Gene Transcripts

Gene Symbol
loxl1
Entrez Gene ID
100124878
Gene Name
lysyl oxidase like 1
Gene Aliases
lol, loxl, loxl-1
Chromosome Location
Chr.3: 82947857 - 82954270 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102831.1 NP_001096301.1 4 - 5 1840 87
GenBank mRNA BC135631.1 - 4 - 5 1840 87

View Full Product Details