Assay Design



Gene Transcripts

Gene Symbol
psmg1
Entrez Gene ID
100124915
Gene Name
proteasome (prosome, macropain) assembly chaperone 1
Gene Aliases
c21lrp, dscr2, lrpc21, pac1
Chromosome Location
Chr.2: 7801172 - 7807435 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102859.1 NP_001096329.1 2 - 3 292 90
GenBank mRNA BC135747.1 - 2 - 3 292 90
CR848288.2 - 2 - 3 239 90

View Full Product Details