Assay Design



Gene Transcripts

Gene Symbol
fktn
Entrez Gene ID
100124956
Gene Name
fukutin
Gene Aliases
-
Chromosome Location
Chr.1: 108100648 - 108114552 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102894.1 NP_001096364.1 2 - 3 243 113
GenBank mRNA BC127298.1 - 2 - 3 264 113
BC135876.1 - 2 - 3 243 113

View Full Product Details