Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102935.1 | NP_001096405.1 | 7 - 8 | 1426 | 94 |
XM_012965901.2 | XP_012821355.1 | 12 - 13 | 3197 | 94 | |
XM_012965902.2 | XP_012821356.1 | 12 - 13 | 3146 | 94 | |
XM_012965903.2 | XP_012821357.1 | 11 - 12 | 3098 | 94 | |
XM_012965904.2 | XP_012821358.1 | 12 - 13 | 3210 | 94 | |
XM_012965906.2 | XP_012821360.1 | 12 - 13 | 1956 | 94 | |
XM_012965908.2 | XP_012821362.1 | 12 - 13 | 1905 | 94 | |
XM_012965909.2 | XP_012821363.1 | 12 - 13 | 1886 | 94 | |
XM_012965910.2 | XP_012821364.1 | 12 - 13 | 1893 | 94 | |
XM_012965911.2 | XP_012821365.1 | 12 - 13 | 1886 | 94 | |
XM_012965912.2 | XP_012821366.1 | 12 - 13 | 1826 | 94 | |
XM_012965913.2 | XP_012821367.1 | 12 - 13 | 1836 | 94 | |
XM_012965914.2 | XP_012821368.1 | 11 - 12 | 1785 | 94 | |
GenBank mRNA | BC136045.1 | - | 7 - 8 | 1426 | 94 |