Assay Design



Gene Transcripts

Gene Symbol
kcnj13
Entrez Gene ID
100125048
Gene Name
potassium channel, inwardly rectifying subfamily J, member 13
Gene Aliases
-
Chromosome Location
Chr.5: 122948229 - 122953536 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102967.1 NP_001096437.1 3 - 4 907 102
GenBank mRNA BC136132.1 - 3 - 4 907 102

View Full Product Details