Assay Design



Gene Transcripts

Gene Symbol
baat
Entrez Gene ID
100125079
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
-
Chromosome Location
Chr.1: 191353272 - 191363248 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102991.1 NP_001096461.1 5 - 6 992 82
GenBank mRNA BC136191.1 - 5 - 6 992 82

View Full Product Details