Assay Design



Gene Transcripts

Gene Symbol
syt17
Entrez Gene ID
100125097
Gene Name
synaptotagmin 17
Gene Aliases
-
Chromosome Location
Chr.9: 26795216 - 26847381 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103008.1 NP_001096478.1 3 - 4 240 68
GenBank mRNA BC136225.1 - 3 - 4 240 68

View Full Product Details