Assay Design



Gene Transcripts

Gene Symbol
nkx6-1
Entrez Gene ID
549337
Gene Name
NK6 homeobox 1
Gene Aliases
nkx6.1, nkx6a
Chromosome Location
Chr.1: 92536067 - 92539344 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016583.3 NP_001016583.2 1 - 2 607 67
GenBank mRNA EF460791.1 - 1 - 2 607 67

View Full Product Details