Assay Design



Gene Transcripts

Gene Symbol
hdgfl3
Entrez Gene ID
100125195
Gene Name
HDGF like 3
Gene Aliases
hdgfrp3
Chromosome Location
Chr.3: 95526477 - 95551459 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103080.1 NP_001096550.1 2 - 3 206 83
XM_012958473.2 XP_012813927.1 2 - 3 607 83
GenBank mRNA BC136239.1 - 2 - 3 206 83

View Full Product Details