Assay Design



Gene Transcripts

Gene Symbol
tsnare1
Entrez Gene ID
100127544
Gene Name
t-SNARE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 131182763 - 131324273 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112923.1 NP_001106394.1 2 - 3 209 86
GenBank mRNA BC152038.1 - 2 - 3 209 86
CT025175.1 - 3 - 4 249 86

View Full Product Details