Assay Design



Gene Transcripts

Gene Symbol
jmy
Entrez Gene ID
100127594
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
-
Chromosome Location
Chr.1: 163889381 - 163914325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112955.1 NP_001106426.1 6 - 7 1516 68
GenBank mRNA BC153370.1 - 6 - 7 1516 68

View Full Product Details