Assay Design



Gene Transcripts

Gene Symbol
prdx5
Entrez Gene ID
100127718
Gene Name
peroxiredoxin 5
Gene Aliases
acr1, aoeb166, pmp20, prx-5, prx5, prxv, sbbi10
Chromosome Location
Chr.4: 39847328 - 39854018 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113054.1 NP_001106525.1 3 - 4 350 69
GenBank mRNA BC154896.1 - 3 - 4 350 69

View Full Product Details