Assay Design



Gene Transcripts

Gene Symbol
cfap52
Entrez Gene ID
100127736
Gene Name
cilia and flagella associated protein 52
Gene Aliases
wdr16, wdrpuh
Chromosome Location
Chr.10: 39011284 - 39016474 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113074.1 NP_001106545.1 3 - 4 429 64
GenBank mRNA BC154922.1 - 3 - 4 429 64

View Full Product Details