Assay Design



Gene Transcripts

Gene Symbol
otop3
Entrez Gene ID
100127796
Gene Name
otopetrin 3
Gene Aliases
Nlo
Chromosome Location
Chr.10: 35665864 - 35675710 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113113.1 NP_001106584.1 2 - 3 588 62
XM_012971513.2 XP_012826967.1 3 - 4 729 62
GenBank mRNA BC155406.1 - 2 - 3 588 62

View Full Product Details