Assay Design



Gene Transcripts

Gene Symbol
enosf1
Entrez Gene ID
100127818
Gene Name
enolase superfamily member 1
Gene Aliases
-
Chromosome Location
Chr.6: 93015201 - 93039954 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113128.1 NP_001106599.1 13 - 14 1025 98
XM_012964234.2 XP_012819688.1 14 - 15 1455 98
XM_012964235.2 XP_012819689.1 14 - 15 1372 98
XM_012964236.2 XP_012819690.1 14 - 15 1346 98
XM_018094523.1 XP_017950012.1 13 - 14 1544 98
GenBank mRNA BC155430.1 - 13 - 14 1025 98

View Full Product Details