Assay Design



Gene Transcripts

Gene Symbol
anos1
Entrez Gene ID
100135017
Gene Name
anosmin 1
Gene Aliases
admlx, hha, kal, kal1, kalig-1, kms
Chromosome Location
Chr.2: 28706249 - 28821040 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113688.1 NP_001107160.1 8 - 9 1392 96
GenBank mRNA BC155677.1 - 8 - 9 1392 96
BC170858.1 - 8 - 9 1267 96

View Full Product Details