Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113817.1 | NP_001107289.1 | 3 - 4 | 427 | 94 |
XM_012960557.2 | XP_012816011.1 | 3 - 4 | 358 | 94 | |
XM_012960558.2 | XP_012816012.1 | 3 - 4 | 357 | 94 | |
GenBank mRNA | BC155683.1 | - | 3 - 4 | 427 | 94 |
BC171146.1 | - | 3 - 4 | 425 | 94 |