Assay Design



Gene Transcripts

Gene Symbol
aftph
Entrez Gene ID
100135100
Gene Name
aftiphilin
Gene Aliases
-
Chromosome Location
Chr.5: 29032949 - 29067478 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113838.1 NP_001107310.1 3 - 4 2132 69
XM_012962672.2 XP_012818126.1 3 - 4 2029 69
XM_012962675.2 XP_012818129.1 3 - 4 2029 69
GenBank mRNA BC155706.1 - 3 - 4 2132 69
CT025421.2 - 3 - 4 2031 69

View Full Product Details