Assay Design



Gene Transcripts

Gene Symbol
clps
Entrez Gene ID
100135140
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.2: 61379374 - 61397635 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113855.1 NP_001107327.1 1 - 2 115 84
GenBank mRNA BC156015.1 - 1 - 2 115 84

View Full Product Details