Assay Design



Gene Transcripts

Gene Symbol
kiaa0930
Entrez Gene ID
100135200
Gene Name
KIAA0930
Gene Aliases
c22orf9
Chromosome Location
Chr.Un|NW_016683675: 10333 - 14364 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113902.1 NP_001107374.1 - 672 90
GenBank mRNA BC157243.1 - - 672 90
BC171214.1 - - 661 90
BC171216.1 - - 661 90

View Full Product Details