Assay Design



Gene Transcripts

Gene Symbol
cndp1
Entrez Gene ID
100135225
Gene Name
carnosine dipeptidase 1 (metallopeptidase M20 family)
Gene Aliases
cn1, cpgl2
Chromosome Location
Chr.Un|NW_016683375: 1572317 - 1585929 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113922.1 NP_001107394.1 10 - 11 1404 87
GenBank mRNA BC157332.1 - 10 - 11 1404 87
BC170995.1 - 10 - 11 1401 87
BC171000.1 - 10 - 11 1401 87

View Full Product Details