Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113922.1 | NP_001107394.1 | 1 - 2 | 114 | 100 |
GenBank mRNA | BC157332.1 | - | 1 - 2 | 114 | 100 |
BC170995.1 | - | 1 - 2 | 111 | 100 | |
BC171000.1 | - | 1 - 2 | 111 | 100 |