Assay Design



Gene Transcripts

Gene Symbol
was
Entrez Gene ID
496673
Gene Name
Wiskott-Aldrich syndrome
Gene Aliases
imd2, thc, thc1, wasp
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114203.1 NP_001107675.1 - 763 90
GenBank mRNA BC087802.1 - - 739 90
BC158507.1 - - 763 90

View Full Product Details