Assay Design



Gene Transcripts

Gene Symbol
zcrb1
Entrez Gene ID
100135752
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Gene Aliases
madp-1, madp1, rbm36, zcchc19
Chromosome Location
Chr.3: 55298369 - 55308640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114501.2 NP_001107973.1 5 - 6 438 95
XM_012958532.2 XP_012813986.1 6 - 7 530 95
GenBank mRNA BC158310.1 - 5 - 6 440 95
CU025042.1 - 6 - 7 465 95

View Full Product Details