Assay Design



Gene Transcripts

Gene Symbol
krt12.5
Entrez Gene ID
100144436
Gene Name
keratin 12, gene 5
Gene Aliases
Krt12.5.S, k12, krt12, xk81b2
Chromosome Location
Chr.Un|NW_016683386: 1343918 - 1347976 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123024.1 NP_001116496.1 6 - 7 1227 102
GenBank mRNA BC158426.1 - 6 - 7 1227 102
BC158447.1 - 6 - 7 1227 102
CU025121.1 - 6 - 7 1192 102

View Full Product Details