Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123422.1 | NP_001116894.1 | 2 - 3 | 242 | 86 |
XM_012967973.2 | XP_012823427.1 | 2 - 3 | 293 | 86 | |
XM_012967974.2 | XP_012823428.1 | 3 - 4 | 407 | 86 | |
GenBank mRNA | BC159321.1 | - | 2 - 3 | 242 | 86 |