Assay Design



Gene Transcripts

Gene Symbol
best4
Entrez Gene ID
100144681
Gene Name
bestrophin 4
Gene Aliases
vmd2l2
Chromosome Location
Chr.4: 72625093 - 72640179 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123442.1 NP_001116914.1 2 - 3 501 70
GenBank mRNA BC161756.1 - 2 - 3 501 70

View Full Product Details